Assay Details
| Gene Symbol: Slc13a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172892.3 | NP_766480.1 | 6 - 7 | 1297 | 82 | |
| GenBank mRNA | AK036354.1 | - | 6 - 7 | 1299 | 82 | |
| AK038937.1 | - | 6 - 7 | 1302 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc13a4
Genomic Map