Assay Details
| Gene Symbol: Nhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081052.2 | NP_001074521.1 | 6 - 7 | 4204 | 71 | |
| NM_001290526.1 | NP_001277455.1 | 7 - 8 | 4267 | 71 | ||
| GenBank mRNA | BC147632.1 | - | 6 - 7 | 4204 | 9056622 | 71 |
| BC158016.1 | - | 7 - 8 | 4267 | 9007322 | 71 | |
| BC172134.1 | - | 7 - 8 | 4267 | 9054804 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nhs
Genomic Map