Assay Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290633.1 | NP_001277562.1 | 11 - 12 | 1441 | 122 | |
| NM_178256.4 | NP_839987.2 | 11 - 12 | 1444 | 122 | ||
| GenBank mRNA | AK147432.1 | - | 8 - 9 | 723 | 122 | |
| AY263368.1 | - | 11 - 12 | 1389 | 122 | ||
| BC139012.1 | - | 11 - 12 | 1320 | 8862034 | 122 | |
| BC145500.1 | - | 11 - 12 | 1317 | 9054063 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reps2
Genomic Map