Assay Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290633.1 | NP_001277562.1 | 13 - 14 | 1624 | 98 | |
| NM_178256.4 | NP_839987.2 | 13 - 14 | 1627 | 98 | ||
| GenBank mRNA | AK147432.1 | - | 10 - 11 | 906 | 98 | |
| AY263368.1 | - | 13 - 14 | 1572 | 98 | ||
| BC139012.1 | - | 13 - 14 | 1503 | 8862034 | 98 | |
| BC145500.1 | - | 13 - 14 | 1500 | 9054063 | 98 | |
More Information
Set Membership: |
|

Reps2
Genomic Map