Assay Details
| Gene Symbol: Reps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290633.1 | NP_001277562.1 | 5 - 6 | 875 | 61 | |
| NM_178256.4 | NP_839987.2 | 5 - 6 | 878 | 61 | ||
| GenBank mRNA | AK147418.1 | - | 5 - 6 | 526 | 61 | |
| AK147432.1 | - | 2 - 3 | 157 | 61 | ||
| AY263368.1 | - | 5 - 6 | 823 | 61 | ||
| BC139012.1 | - | 5 - 6 | 754 | 8862034 | 61 | |
| BC145500.1 | - | 5 - 6 | 751 | 9054063 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reps2
Genomic Map