Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004361.2 | NP_001004361.2 | 1 - 2 | 354 | 80 | |
| NM_001142698.1 | NP_001136170.1 | 1 - 2 | 354 | 80 | ||
| GenBank mRNA | AK017728.1 | - | 1 - 2 | 108 | 80 | |
| AK141632.1 | - | 1 - 2 | 172 | 80 | ||
| BC080777.1 | - | 1 - 2 | 110 | 30439612 | 80 | |
| BQ770487.1 | - | 1 - 2 | 354 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC080777.1 transcripts.
Set Membership: |
|

Tpgs2
Genomic Map