Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004361.2 | NP_001004361.2 | 6 - 7 | 922 | 82 | |
| NM_001142697.1 | NP_001136169.1 | 6 - 7 | 668 | 82 | ||
| GenBank mRNA | AK160220.1 | - | 6 - 7 | 718 | 82 | |
| AK178089.1 | - | 1 - 2 | 82 | |||
| BC060253.1 | - | 4 - 5 | 474 | 82 | ||
| BC080777.1 | - | 6 - 7 | 678 | 30439612 | 82 | |
| DV655125.1 | - | 6 - 7 | 83 | |||
More Information
Set Membership: |
|

Tpgs2
Genomic Map