Assay Details
| Gene Symbol: Mocos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026779.1 | NP_081055.1 | 6 - 7 | 1447 | 68 | |
| GenBank mRNA | AK076385.1 | - | 6 - 7 | 1261 | 68 | |
| BC016223.1 | - | 6 - 7 | 1262 | 68 | ||
| BC113180.1 | - | 6 - 7 | 1447 | 40060723 | 68 | |
| BC113786.1 | - | 6 - 7 | 1447 | 40060721 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mocos
Genomic Map