Assay Details
| Gene Symbol: Strip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037740.1 | NP_001032829.1 | 13 - 14 | 1566 | 73 | |
| NM_177204.3 | NP_796178.2 | 13 - 14 | 1566 | 73 | ||
| NR_104576.1 | 13 - 14 | 1562 | 73 | |||
| GenBank mRNA | AK041004.1 | - | 13 - 14 | 1567 | 73 | |
| AK042073.1 | - | 13 - 14 | 1567 | 73 | ||
| AK052271.1 | - | 12 - 13 | 1974 | 73 | ||
| AK052273.1 | - | 5 - 6 | 591 | 73 | ||
| AK122459.1 | - | 13 - 14 | 1371 | 73 | ||
| AK163706.1 | - | 13 - 14 | 1566 | 73 | ||
| BC147502.1 | - | 13 - 14 | 1558 | 9056385 | 73 | |
| BC172034.1 | - | 13 - 14 | 1554 | 73 | ||
More Information
Set Membership: |
|

Strip2
Genomic Map