| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
TC0600000250.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
NM_001037740.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
NM_001037740.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
NM_177204.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
NM_177204.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001773.mm.1 |
NR_104576.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
TC0600000250.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
NM_001037740.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
NM_001037740.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
NM_177204.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
NM_177204.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001772.mm.1 |
NR_104576.1 |
Refseq |