Assay Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177676.6 | NP_808344.3 | 11 - 12 | 1498 | 111 | |
| NR_033188.1 | 10 - 11 | 1988 | 111 | |||
| GenBank mRNA | AK029839.1 | - | 10 - 11 | 1930 | 111 | |
| BC137999.1 | - | 7 - 8 | 1219 | 8861018 | 111 | |
| BC145792.1 | - | 7 - 8 | 1221 | 40131095 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

4931409K22Rik
Genomic Map