Assay Details
| Gene Symbol: 4931409K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177676.6 | NP_808344.3 | 13 - 14 | 1644 | 80 | |
| NR_033188.1 | 12 - 13 | 2134 | 80 | |||
| GenBank mRNA | AK029839.1 | - | 12 - 13 | 2076 | 80 | |
| BC137999.1 | - | 9 - 10 | 1365 | 8861018 | 80 | |
| BC145792.1 | - | 9 - 10 | 1367 | 40131095 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

4931409K22Rik
Genomic Map