Assay Details
| Gene Symbol: Atg9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002897.3 | NP_001002897.2 | 7 - 8 | 2102 | 88 | |
| GenBank mRNA | AY495541.1 | - | 7 - 8 | 2102 | 88 | |
| AY515312.1 | - | 2 - 3 | 275 | 88 | ||
| AY515313.1 | - | 7 - 8 | 2102 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg9b
Genomic Map