Assay Details
| Gene Symbol: Atg9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002897.3 | NP_001002897.2 | 8 - 9 | 2281 | 119 | |
| GenBank mRNA | AY495541.1 | - | 8 - 9 | 2281 | 119 | |
| AY515312.1 | - | 3 - 4 | 454 | 119 | ||
| AY515313.1 | - | 8 - 9 | 2281 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541452 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541451 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183899 | AY515313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183899 | AY515312.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183899 | NM_001002897.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183899 | AK158106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183899 | AW489695.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19936 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132711 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143283 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183899 | AY515313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183899 | AY515312.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183899 | NM_001002897.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183899 | AK158106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183899 | AW489695.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541486 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05506787 | - | - |
| Whole Mouse Genome G4122A | A_52_P430369 | AY515313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P430369 | AY515312.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P430369 | AY495541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P430369 | NM_001002897.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116937 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04159264 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04234121 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04234114 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04503798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04503807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04234111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04503806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04234119 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04503800 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04234116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04503808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04150522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541451 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541452 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541455 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04234119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04234120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04503809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04866770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04234125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04503807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04503808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04575525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528579 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528580 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528573 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528575 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528574 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528578 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434092_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445674_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528567 | - | - |
| Mouse Genome 430 2.0 Array | 1434092_at | - | - |
| Mouse Genome 430 2.0 Array | 1444909_at | - | - |
| Mouse Genome 430 2.0 Array | 1444909_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445674_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434092_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445674_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213948_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213948_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500018857.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018857.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018857.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018859.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018859.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018859.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018851.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018851.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018851.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018858.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018858.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018858.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018856.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018856.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018856.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018850.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018850.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018850.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018852.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018852.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018852.mm.1 | NM_001002897.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018854.mm.1 | TC0500002075.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018854.mm.1 | NM_001002897.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018854.mm.1 | NM_001002897.3 | Refseq |
| Mouse Expression Set 430 | 1434092_PM_at | - | - |
| Mouse Expression Set 430 | 1434092_at | - | - |
| Mouse Expression Set 430 | 1445674_at | - | - |
| Mouse Expression Set 430 | 1445674_PM_at | - | - |
| Murine Genome U74v2 Set | 136639_at | - | - |
| Murine Genome U74v2 Set | 136639_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434092_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434092_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434092_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434092_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445674_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445674_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 537254 | - | - |
| ABI Mouse Genome Survey Microarray | 765315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160369 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3160369 | - | - |
| mouse-6 v1.1 (Illumina) | GI_51890224-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3160369 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_51890224-S | - | - |

Atg9b
Genomic Map