Assay Details
| Gene Symbol: Reep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007874.3 | NP_031900.3 | 1 - 2 | 211 | 71 | |
| GenBank mRNA | AK010507.1 | - | 1 - 2 | 163 | 71 | |
| AK013146.1 | - | 1 - 2 | 142 | 71 | ||
| AK151058.1 | - | 1 - 2 | 166 | 71 | ||
| AK151271.1 | - | 1 - 2 | 166 | 71 | ||
| AK152361.1 | - | 1 - 2 | 140 | 71 | ||
| AK152503.1 | - | 1 - 2 | 166 | 71 | ||
| AK167385.1 | - | 1 - 2 | 140 | 71 | ||
| AY562233.1 | - | 1 - 2 | 117 | 71 | ||
| BC013052.1 | - | 1 - 2 | 142 | 3585290 | 71 | |
| BU053115.1 | - | 1 - 2 | 211 | 71 | ||
| U28168.1 | - | 1 - 2 | 140 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Reep5
Genomic Map