Assay Details
| Gene Symbol: D630045J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194061.2 | NP_918950.2 | 18 - 19 | 5425 | 57 | |
| GenBank mRNA | AK085604.1 | - | 4 - 5 | 786 | 57 | |
| AK128948.1 | - | 4 - 5 | 839 | 57 | ||
| BC079889.1 | - | 17 - 18 | 4787 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

D630045J12Rik
Genomic Map