Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012031.3 | NP_036161.2 | 13 - 14 | 1884 | 65 | |
| GenBank mRNA | AF181252.1 | - | 13 - 14 | 1876 | 65 | |
| AF181253.1 | - | 3 - 4 | 568 | 65 | ||
| AK032601.1 | - | 10 - 11 | 1679 | 65 | ||
| BC046313.1 | - | 13 - 14 | 1884 | 6310604 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag1
Genomic Map