Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012031.3 | NP_036161.2 | 15 - 16 | 2169 | 143 | |
| GenBank mRNA | AF181252.1 | - | 15 - 16 | 2161 | 143 | |
| AF181253.1 | - | 5 - 6 | 853 | 143 | ||
| AK032601.1 | - | 12 - 13 | 1964 | 143 | ||
| BC046313.1 | - | 15 - 16 | 2169 | 6310604 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag1
Genomic Map