| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500001945.mm.1 |
NM_012031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001945.mm.1 |
NM_012031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001945.mm.1 |
TC1500000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001946.mm.1 |
NM_012031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001946.mm.1 |
NM_012031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001946.mm.1 |
TC1500000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001944.mm.1 |
NM_012031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001944.mm.1 |
NM_012031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001944.mm.1 |
TC1500000263.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500001947.mm.1 |
NM_012031.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001947.mm.1 |
NM_012031.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500001947.mm.1 |
TC1500000263.mm.1 |
MTA TC |