Assay Details
| Gene Symbol: Spag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012031.3 | NP_036161.2 | 4 - 5 | 665 | 109 | |
| GenBank mRNA | AF181252.1 | - | 4 - 5 | 657 | 109 | |
| BC046313.1 | - | 4 - 5 | 665 | 6310604 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag1
Genomic Map