Assay Details
| Gene Symbol: Rnf19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013923.2 | NP_038951.1 | 1 - 2 | 237 | 119 | |
| GenBank mRNA | AK082665.1 | - | 1 - 2 | 189 | 119 | |
| AK147768.1 | - | 1 - 2 | 225 | 119 | ||
| AK166197.1 | - | 1 - 2 | 225 | 119 | ||
| BY207861.1 | - | 1 - 2 | 238 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK147768.1, AK166197.1, NM_013923.2 transcripts.
Set Membership: |
|

Rnf19a
Genomic Map