Assay Details
| Gene Symbol: Paip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026420.2 | NP_080696.1 | 2 - 3 | 252 | 121 | |
| GenBank mRNA | AK005462.1 | - | 2 - 3 | 254 | 121 | |
| AK009909.1 | - | 2 - 3 | 253 | 121 | ||
| AK146327.1 | - | 2 - 3 | 283 | 121 | ||
| AK150957.1 | - | 2 - 3 | 260 | 121 | ||
| AK154453.1 | - | 3 - 4 | 354 | 121 | ||
| AK167545.1 | - | 2 - 3 | 279 | 121 | ||
| AK169207.1 | - | 2 - 3 | 274 | 121 | ||
| AK169411.1 | - | 2 - 3 | 274 | 121 | ||
| BC010190.1 | - | 2 - 3 | 268 | 3964799 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paip2
Genomic Map