Assay Details
| Gene Symbol: Psd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289602.1 | NP_001276531.1 | 3 - 4 | 1247 | 61 | |
| NM_028707.4 | NP_082983.3 | 3 - 4 | 1244 | 61 | ||
| NR_110349.1 | 3 - 4 | 1244 | 61 | |||
| GenBank mRNA | AK034575.1 | - | 3 - 4 | 1010 | 61 | |
| AK045529.1 | - | 3 - 4 | 972 | 61 | ||
| AK053835.1 | - | 3 - 4 | 972 | 61 | ||
| AK149027.1 | - | 3 - 4 | 971 | 61 | ||
| BC062930.1 | - | 2 - 3 | 825 | 6855293 | 61 | |
| BC065053.1 | - | 3 - 4 | 1009 | 5709231 | 61 | |
| BC065055.1 | - | 3 - 4 | 995 | 5720949 | 61 | |
| BC066026.1 | - | 3 - 4 | 1244 | 5721455 | 61 | |
| BC066036.1 | - | 3 - 4 | 1010 | 6837266 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd2
Genomic Map