Assay Details
| Gene Symbol: Nrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167891.1 | NP_001161363.1 | 7 - 8 | 1445 | 112 | |
| GenBank mRNA | AW476657.1 | - | 5 - 6 | 486 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nrg2
Genomic Map