Assay Details
| Gene Symbol: Ecscr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033141.1 | NP_001028313.1 | 2 - 3 | 213 | 66 | |
| GenBank mRNA | AK157485.1 | - | 2 - 3 | 213 | 66 | |
| EU025067.1 | - | 2 - 3 | 149 | 66 | ||
More Information
Set Membership: |
|

Ecscr
Genomic Map