Assay Details
| Gene Symbol: Ecscr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033141.1 | NP_001028313.1 | 5 - 6 | 535 | 70 | |
| GenBank mRNA | AK157485.1 | - | 5 - 6 | 535 | 70 | |
| EU025067.1 | - | 5 - 6 | 471 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ecscr
Genomic Map