Assay Details
| Gene Symbol: Tmem173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289591.1 | NP_001276520.1 | 5 - 6 | 1008 | 62 | |
| NM_001289592.1 | NP_001276521.1 | 6 - 7 | 975 | 62 | ||
| NM_028261.1 | NP_082537.1 | 6 - 7 | 1098 | 62 | ||
| GenBank mRNA | AK012006.1 | - | 6 - 7 | 1045 | 62 | |
| AK077788.1 | - | 6 - 7 | 999 | 62 | ||
| AK089405.1 | - | 6 - 7 | 1076 | 62 | ||
| AK146284.1 | - | 6 - 7 | 971 | 62 | ||
| AK153868.1 | - | 6 - 7 | 969 | 62 | ||
| AK157370.1 | - | 5 - 6 | 910 | 62 | ||
| AK158458.1 | - | 6 - 7 | 884 | 62 | ||
| AK170724.1 | - | 6 - 7 | 1076 | 62 | ||
| AK171065.1 | - | 6 - 7 | 1062 | 62 | ||
| AK171203.1 | - | 6 - 7 | 1062 | 62 | ||
| AK171612.1 | - | 6 - 7 | 1029 | 62 | ||
| AX775911.1 | - | 6 - 7 | 985 | 62 | ||
| BC027757.1 | - | 4 - 5 | 742 | 62 | ||
| BC046640.1 | - | 5 - 6 | 933 | 5067343 | 62 | |
| DQ910493.1 | - | 6 - 7 | 896 | 62 | ||
| FJ222242.1 | - | 4 - 5 | 760 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem173
Genomic Map