Assay Details
| Gene Symbol: Odf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008757.3 | NP_032783.2 | 1 - 2 | 504 | 88 | |
| GenBank mRNA | BC145758.1 | - | 1 - 2 | 504 | 40131061 | 88 |
| CJ042343.1 | - | 1 - 2 | 393 | 88 | ||
| EU791469.1 | - | 1 - 2 | 504 | 88 | ||
| X79446.1 | - | 1 - 2 | 508 | 88 | ||
More Information
Set Membership: |
|

Odf1
Genomic Map