Assay Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025494.3 | NP_079770.2 | 10 - 11 | 1041 | 81 | |
| GenBank mRNA | AB088407.1 | - | 10 - 11 | 988 | 81 | |
| AK075779.1 | - | 10 - 11 | 1013 | 81 | ||
| AK129007.1 | - | 8 - 9 | 2563 | 81 | ||
| AK151237.1 | - | 10 - 11 | 1002 | 81 | ||
| AK168914.1 | - | 10 - 11 | 992 | 81 | ||
| BC010217.1 | - | 10 - 11 | 989 | 3498388 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c1
Genomic Map