Assay Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025494.3 | NP_079770.2 | 5 - 6 | 593 | 108 | |
| GenBank mRNA | AB088407.1 | - | 5 - 6 | 540 | 108 | |
| AK006323.1 | - | 5 - 6 | 544 | 108 | ||
| AK075779.1 | - | 5 - 6 | 565 | 108 | ||
| AK151237.1 | - | 5 - 6 | 554 | 108 | ||
| AK168914.1 | - | 5 - 6 | 544 | 108 | ||
| BC010217.1 | - | 5 - 6 | 541 | 3498388 | 108 | |
| U13839.1 | - | 5 - 6 | 536 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c1
Genomic Map