Assay Details
| Gene Symbol: Atp6v1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025494.3 | NP_079770.2 | 7 - 8 | 788 | 69 | |
| GenBank mRNA | AB088407.1 | - | 7 - 8 | 735 | 69 | |
| AK075779.1 | - | 7 - 8 | 760 | 69 | ||
| AK129007.1 | - | 5 - 6 | 2310 | 69 | ||
| AK151237.1 | - | 7 - 8 | 749 | 69 | ||
| AK168914.1 | - | 7 - 8 | 739 | 69 | ||
| AK194104.1 | - | 1 - 2 | 77 | 69 | ||
| BC010217.1 | - | 7 - 8 | 736 | 3498388 | 69 | |
| U13839.1 | - | 7 - 8 | 731 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1c1
Genomic Map