| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800001177.mm.1 |
NM_008586.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001177.mm.1 |
NM_008586.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001177.mm.1 |
TC1800000165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001175.mm.1 |
NM_008586.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001175.mm.1 |
NM_008586.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001175.mm.1 |
TC1800000165.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001178.mm.1 |
NM_008586.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001178.mm.1 |
NM_008586.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001178.mm.1 |
TC1800000165.mm.1 |
MTA TC |