Assay Details
| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019737.2 | NP_062711.1 | 2 - 3 | 1167 | 111 | |
| GenBank mRNA | AF097158.1 | - | 2 - 3 | 1165 | 111 | |
| AF142674.1 | - | 2 - 3 | 375 | 111 | ||
| AK137385.1 | - | 2 - 3 | 1147 | 111 | ||
| AK138247.1 | - | 2 - 3 | 1065 | 111 | ||
| AK138610.1 | - | 2 - 3 | 1149 | 111 | ||
| AK144997.1 | - | 2 - 3 | 1041 | 111 | ||
| AK163956.1 | - | 2 - 3 | 1106 | 111 | ||
| AK164262.1 | - | 2 - 3 | 1106 | 111 | ||
| AK164724.1 | - | 2 - 3 | 1106 | 111 | ||
| BC011149.1 | - | 2 - 3 | 390 | 4037384 | 111 | |
More Information
Set Membership: |
|

B4galt6
Genomic Map