Assay Details
| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019737.2 | NP_062711.1 | 7 - 8 | 1835 | 93 | |
| GenBank mRNA | AF097158.1 | - | 7 - 8 | 1833 | 93 | |
| AF142674.1 | - | 7 - 8 | 1043 | 93 | ||
| AK137385.1 | - | 7 - 8 | 1815 | 93 | ||
| AK138247.1 | - | 7 - 8 | 1733 | 93 | ||
| AK138610.1 | - | 7 - 8 | 1817 | 93 | ||
| AK144997.1 | - | 7 - 8 | 1709 | 93 | ||
| AK163956.1 | - | 7 - 8 | 1774 | 93 | ||
| AK164262.1 | - | 7 - 8 | 1774 | 93 | ||
| AK164724.1 | - | 7 - 8 | 1774 | 93 | ||
| BC011149.1 | - | 7 - 8 | 1058 | 4037384 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

B4galt6
Genomic Map