Assay Details
| Gene Symbol: Trappc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029491.2 | NP_083767.2 | 9 - 10 | 1401 | 135 | |
| NM_177038.3 | NP_796012.2 | 9 - 10 | 1401 | 135 | ||
| GenBank mRNA | AK083747.1 | - | 9 - 10 | 1401 | 135 | |
| AK173085.1 | - | 2 - 3 | 178 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trappc8
Genomic Map