Assay Details
| Gene Symbol: Garem | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033445.2 | NP_001028617.2 | 4 - 5 | 1935 | 55 | |
| GenBank mRNA | AK148318.1 | - | 4 - 5 | 1936 | 55 | |
| AK162177.1 | - | 1 - 2 | 60 | 55 | ||
| AK220564.1 | - | 4 - 5 | 1919 | 55 | ||
| BC049983.1 | - | 1 - 2 | 913 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Garem
Genomic Map