| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800002237.mm.1 |
NM_009818.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002237.mm.1 |
NM_009818.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002237.mm.1 |
TC1800000306.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002235.mm.1 |
NM_009818.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002235.mm.1 |
NM_009818.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002235.mm.1 |
TC1800000306.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002236.mm.1 |
NM_009818.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002236.mm.1 |
NM_009818.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002236.mm.1 |
TC1800000306.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800002238.mm.1 |
NM_009818.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002238.mm.1 |
NM_009818.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800002238.mm.1 |
TC1800000306.mm.1 |
MTA TC |