Assay Details
| Gene Symbol: Kif20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166406.1 | NP_001159878.1 | 15 - 16 | 2174 | 92 | |
| NM_001166407.1 | NP_001159879.1 | 15 - 16 | 2172 | 92 | ||
| NM_009004.4 | NP_033030.1 | 15 - 16 | 2068 | 92 | ||
| GenBank mRNA | AK051201.1 | - | 15 - 16 | 1986 | 92 | |
| AK051995.1 | - | 15 - 16 | 2023 | 92 | ||
| AK053280.1 | - | 15 - 16 | 1987 | 92 | ||
| AK083412.1 | - | 15 - 16 | 2028 | 92 | ||
| AK083697.1 | - | 15 - 16 | 1989 | 92 | ||
| AK084732.1 | - | 15 - 16 | 2010 | 92 | ||
| AK142764.1 | - | 15 - 16 | 2016 | 92 | ||
| AK144844.1 | - | 15 - 16 | 1986 | 92 | ||
| BC060608.1 | - | 15 - 16 | 2174 | 5720408 | 92 | |
| Y09632.1 | - | 15 - 16 | 2223 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kif20a
Genomic Map