Assay Details
| Gene Symbol: Hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010481.2 | NP_034611.2 | 1 - 2 | 183 | 133 | |
| GenBank mRNA | AK002634.1 | - | 1 - 2 | 115 | 133 | |
| AK004946.1 | - | 1 - 2 | 145 | 133 | ||
| AK030299.1 | - | 1 - 2 | 176 | 133 | ||
| AK133501.1 | - | 1 - 2 | 185 | 133 | ||
| AK137109.1 | - | 1 - 2 | 155 | 133 | ||
| AK145965.1 | - | 1 - 2 | 177 | 133 | ||
| AK159791.1 | - | 1 - 2 | 185 | 133 | ||
| AK161587.1 | - | 1 - 2 | 185 | 133 | ||
| AK165958.1 | - | 1 - 2 | 183 | 133 | ||
| AK167856.1 | - | 1 - 2 | 176 | 133 | ||
| BC052727.1 | - | 1 - 2 | 183 | 6813991 | 133 | |
| BC057343.1 | - | 1 - 2 | 183 | 6813991 | 133 | |
| BY146974.1 | - | 1 - 2 | 189 | 133 | ||
| D11089.2 | - | 1 - 2 | 171 | 133 | ||
| D17556.1 | - | 1 - 2 | 155 | 133 | ||
| L06896.1 | - | 1 - 2 | 168 | 133 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002634.1, AK004946.1, AK133501.1, AK137109.1, AK145965.1, AK159791.1, AK165958.1, AK167856.1, BC052727.1, BC057343.1, D11089.2, D17556.1, NM_010481.2 transcripts.
Set Membership: |
|

Hspa9
Genomic Map