Assay Details
| Gene Symbol: Snhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038073.1 | 1 - 2 | 63 | 74 | ||
| GenBank mRNA | AK041908.1 | - | 1 - 2 | 64 | 74 | |
| AK088959.1 | - | 1 - 2 | 61 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snhg4
Genomic Map