Assay Details
| Gene Symbol: Cped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081351.1 | NP_001074820.1 | 12 - 13 | 2531 | 104 | |
| GenBank mRNA | AK032959.1 | - | 11 - 12 | 1218 | 104 | |
| BC151018.1 | - | 12 - 13 | 2046 | 9087931 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cped1
Genomic Map