Assay Details
| Gene Symbol: Ppm1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110218.1 | NP_001103688.1 | 9 - 10 | 1814 | 94 | |
| GenBank mRNA | AK051434.1 | - | 9 - 10 | 1773 | 94 | |
| AK134804.1 | - | 9 - 10 | 1794 | 94 | ||
| AK150309.1 | - | 9 - 10 | 1376 | 94 | ||
| AK220260.1 | - | 9 - 10 | 1771 | 94 | ||
| BC052910.1 | - | 9 - 10 | 1125 | 94 | ||
More Information
Set Membership: |
|

Ppm1h
Genomic Map