Assay Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027256.2 | NP_081532.1 | 12 - 13 | 1658 | 120 | |
| GenBank mRNA | BC013710.1 | - | 12 - 13 | 1604 | 3156446 | 120 |
| BC021838.1 | - | 5 - 6 | 661 | 120 | ||
| BC031145.1 | - | 6 - 7 | 737 | 120 | ||
| CF724981.1 | - | 3 - 4 | 445 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ints4
Genomic Map