Assay Details
| Gene Symbol: Tcaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146174.1 | NP_666286.1 | 4 - 5 | 1887 | 89 | |
| GenBank mRNA | AK131169.1 | - | 4 - 5 | 2100 | 89 | |
| BC008555.1 | - | 4 - 5 | 1904 | 3592166 | 89 | |
| BC011487.1 | - | 4 - 5 | 1887 | 3499593 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcaf2
Genomic Map