Assay Details
| Gene Symbol: Arhgef5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133674.1 | NP_598435.1 | 1 - 2 | 102 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133674.1 transcripts.
Set Membership: |
|

Arhgef5
Genomic Map