Assay Details
| Gene Symbol: Sra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164406.1 | NP_001157878.1 | 1 - 2 | 66 | 59 | |
| NM_025291.3 | NP_079567.2 | 1 - 2 | 66 | 59 | ||
| GenBank mRNA | AK010375.1 | - | 1 - 2 | 52 | 59 | |
| AK136702.1 | - | 1 - 2 | 34 | 59 | ||
| AK154232.1 | - | 1 - 2 | 36 | 59 | ||
| BC048362.1 | - | 1 - 2 | 38 | 2646489 | 59 | |
| BY097874.1 | - | 1 - 2 | 66 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010375.1, AK154232.1, BC048362.1 transcripts.
Set Membership: |
|

Sra1
Genomic Map