Assay Details
| Gene Symbol: Ik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011879.2 | NP_036009.1 | 1 - 2 | 131 | 64 | |
| GenBank mRNA | AK029644.1 | - | 1 - 2 | 85 | 64 | |
| AK050625.1 | - | 1 - 2 | 89 | 64 | ||
| AK082356.1 | - | 1 - 2 | 63 | 64 | ||
| AK087976.1 | - | 1 - 2 | 77 | 64 | ||
| AK088908.1 | - | 1 - 2 | 66 | 64 | ||
| AK163766.1 | - | 1 - 2 | 85 | 64 | ||
| AK167238.1 | - | 1 - 2 | 96 | 64 | ||
| BC006706.1 | - | 1 - 2 | 109 | 64 | ||
| BC014739.1 | - | 1 - 2 | 62 | 4920184 | 64 | |
| BC026569.1 | - | 1 - 2 | 62 | 4982612 | 64 | |
| BC029148.1 | - | 1 - 2 | 104 | 64 | ||
| BC029671.1 | - | 1 - 2 | 74 | 5364953 | 64 | |
| BC029694.1 | - | 1 - 2 | 71 | 5369477 | 64 | |
| BC033356.1 | - | 1 - 2 | 58 | 5366230 | 64 | |
| BG084120.2 | - | 1 - 2 | 131 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029644.1, AK050625.1, AK087976.1, AK163766.1, AK167238.1, BC014739.1, BC026569.1, BC029671.1, BC029694.1, BC033356.1, NM_011879.2 transcripts.
Set Membership: |
|

Ik
Genomic Map