Assay Details
| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171634.1 | - | 2 - 3 | 489 | 95 | |
| BC024136.1 | - | 2 - 3 | 294 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apbb3| Gene Symbol: Apbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK171634.1 | - | 2 - 3 | 489 | 95 | |
| BC024136.1 | - | 2 - 3 | 294 | 95 | ||
Set Membership: |
|