Assay Details
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177697.3 | NP_808365.2 | 30 - 31 | 3642 | 69 | |
| GenBank mRNA | AK086942.1 | - | 29 - 30 | 3240 | 69 | |
| AK136898.1 | - | 29 - 30 | 3241 | 69 | ||
More Information
Set Membership: |
|

Vwa3a
Genomic Map