| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800001817.mm.1 |
NM_011946.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001817.mm.1 |
NM_011946.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001817.mm.1 |
TC1800000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001813.mm.1 |
NM_011946.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001813.mm.1 |
NM_011946.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001813.mm.1 |
TC1800000257.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001815.mm.1 |
NM_011946.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001815.mm.1 |
NM_011946.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001815.mm.1 |
TC1800000257.mm.1 |
MTA TC |